Assay Details
Target Gene Details
Entrez Gene ID: | 387836 |
Gene Name: | C-type lectin domain family 2 member A |
Gene Aliases: |
INPE5792, KACL, PILAR, UNQ5792 |
Location: |
Chr.12:9882733-9932430 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLEC2A | NM_001130711.1 | NP_001124183.1 | ||
| NM_207375.2 | NP_997258.1 | |||
| XM_011520654.2 | XP_011518956.1 | |||
| XM_011520656.2 | XP_011518958.1 | |||
| XM_011520657.2 | XP_011518959.1 | |||
| AY359126.1 | AAQ89483.1 | |||
| EF127467.1 | ||||
| EU095393.1 | ||||
| EU095394.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1051530 | Chr.12:8442286 - 9951690 on Build GRCh38 | Gain |
|
| nsv1044587 | Chr.12:9876915 - 9970815 on Build GRCh38 | Loss |
|
| nsv523050 | Chr.12:9879281 - 9971774 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map