Assay Details
Target Gene Details
Entrez Gene ID: | 23067 |
Gene Name: | SET domain containing 1B |
Gene Aliases: |
KMT2G, Set1B |
Location: |
Chr.12:121803665-121832656 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SETD1B | NM_015048.1 | 5 | 1536 | NP_055863.1 |
| XM_005253858.4 | 6 | 1695 | XP_005253915.1 | |
| XM_006719296.3 | 6 | 1611 | XP_006719359.1 | |
| JF813787.1 | 5 | 1530 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160065 | Chr.12:121780583 - 121859371 on Build GRCh38 | Deletion |
|
| esv3892268 | Chr.12:121747658 - 121845875 on Build GRCh38 | Loss |
|
| nsv911 | Chr.12:121796618 - 121841519 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map