Assay Details
Target Gene Details
Entrez Gene ID: | 121355 |
Gene Name: | gametocyte specific factor 1 |
Gene Aliases: |
FAM112B |
Location: |
Chr.12:54455952-54473602 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GTSF1 | NM_144594.2 | NP_653195.2 | ||
| XM_017018800.1 | XP_016874289.1 | |||
| AK057504.1 | 1 | 347 | ||
| AK098819.1 | BAC05423.1 | |||
| BC021179.1 | AAH21179.1 | |||
| BC105587.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508674 | Chr.12:54446126 - 54522212 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map