Assay Details
Target Gene Details
Entrez Gene ID: | 57701 |
Gene Name: | NCK associated protein 5 like |
Gene Aliases: |
Cep169, FP1193, KIAA1602 |
Location: |
Chr.12:49791139-49828750 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCKAP5L | NM_001037806.3 | 13 | 4858 | NP_001032895.2 |
| XM_006719525.2 | 13 | 5153 | XP_006719588.1 | |
| XM_011538610.2 | 14 | 4937 | XP_011536912.1 | |
| XM_011538611.2 | 14 | 4907 | XP_011536913.1 | |
| AB046822.1 | 5 | 4193 | BAB13428.1 | |
| AF370366.1 | 6 | 2161 | AAQ15202.1 | |
| BC007998.1 | 8 | 2251 | AAH07998.1 | |
| BC033253.1 | 6 | 3374 | AAH33253.1 | |
| BU617503.1 | 1 | 61 | ||
| BX327000.2 | 1 | 598 |
More Information
Set Membership: |
|

Genomic Map