Assay Details
Target Gene Details
Entrez Gene ID: | 283450 |
Gene Name: | HECT domain E3 ubiquitin protein ligase 4 |
Gene Aliases: |
C12orf51, POTAGE |
Location: |
Chr.12:112160188-112382092 on Build GRCh38 |
Assay Gene Location: | Within Exon 76 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HECTD4 | NM_001109662.3 | 76 | 15265 | NP_001103132.3 |
| AB014514.3 | 49 | 11035 | BAA31589.3 | |
| AK001372.1 | 3 | 2433 | ||
| AL117469.1 | 2 | 2366 | CAB55944.1 | |
| BC006270.2 | 5 | 2840 | AAH06270.2 | |
| BC021144.1 | 14 | 4237 | AAH21144.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv483114 | Chr.12:112066462 - 112243129 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map