Assay Details
Target Gene Details
Entrez Gene ID: | 339 |
Gene Name: | apolipoprotein B mRNA editing enzyme catalytic subunit 1 |
Gene Aliases: |
APOBEC-1, BEDP, CDAR1, HEPR |
Location: |
Chr.12:7649400-7670599 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APOBEC1 | NM_001304566.1 | NP_001291495.1 | ||
| NM_001644.4 | NP_001635.2 | |||
| NM_005889.3 | NP_005880.2 | |||
| BC096158.1 | AAH96158.1 | |||
| BC101972.1 | AAI01973.1 | |||
| BC101973.1 | AAI01974.1 | |||
| L25877.1 | AAA86766.1 | |||
| L26234.1 | AAA64230.1 | |||
| U72890.1 | AAD09567.1 | |||
| U72891.1 | AAD00185.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1350n100 | Chr.12:7502541 - 7672399 on Build GRCh38 | Loss |
|
| nsv826213 | Chr.12:7578336 - 7678048 on Build GRCh38 | Gain |
|
| nsv1036408 | Chr.12:7630029 - 7659414 on Build GRCh38 | Loss |
|
| nsv826216 | Chr.12:7615958 - 7665910 on Build GRCh38 | Gain |
|
| nsv557273 | Chr.12:7621432 - 7731755 on Build GRCh38 | Loss |
|
| nsv557272 | Chr.12:7604110 - 7669096 on Build GRCh38 | Loss |
|
| dgv5e196 | Chr.12:7621924 - 7954221 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map