Assay Details
Target Gene Details
Entrez Gene ID: | 144481 |
Gene Name: | SOCS2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.12:93565628-93571398 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOCS2-AS1 | NR_038263.1 | |||
| AK054607.1 | ||||
| AK098304.1 |

Genomic Map