Assay Details
Target Gene Details
Entrez Gene ID: | 91523 |
Gene Name: | PC-esterase domain containing 1B |
Gene Aliases: |
FAM113B |
Location: |
Chr.12:47079603-47236663 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCED1B | NM_001281429.1 | NP_001268358.1 | ||
| NM_138371.2 | NP_612380.1 | |||
| XM_005269224.4 | XP_005269281.1 | |||
| XM_011538978.2 | XP_011537280.1 | |||
| XM_017020207.1 | XP_016875696.1 | |||
| XM_017020208.1 | XP_016875697.1 | |||
| XM_017020209.1 | XP_016875698.1 | |||
| XM_017020210.1 | XP_016875699.1 | |||
| XM_017020213.1 | XP_016875702.1 | |||
| XM_017020214.1 | XP_016875703.1 | |||
| XM_017020215.1 | XP_016875704.1 | |||
| XM_017020216.1 | XP_016875705.1 | |||
| CR985281.1 |
Target Gene Details
Entrez Gene ID: | 100233209 |
Gene Name: | PCED1B antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.12:47208420-47216443 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCED1B-AS1 | NR_026544.1 | 3 | 604 | |
| AK123977.1 | 3 | 596 | ||
| DA925061.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832399 | Chr.12:47109127 - 47295818 on Build GRCh38 | Gain |
|
| nsv558784 | Chr.12:46981125 - 47281897 on Build GRCh38 | Gain |
|
| esv2751100 | Chr.12:47147607 - 47352374 on Build GRCh38 | Loss |
|
| esv3580259 | Chr.12:47206711 - 47218023 on Build GRCh38 | Loss |
|
| esv3892198 | Chr.12:47149024 - 47608617 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map