Assay Details
Target Gene Details
Entrez Gene ID: | 80018 |
Gene Name: | N(alpha)-acetyltransferase 25, NatB auxiliary subunit |
Gene Aliases: |
C12orf30, MDM20, NAP1 |
Location: |
Chr.12:112026689-112109022 on Build GRCh38 |
Assay Gene Location: | Within Intron 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAA25 | NM_024953.3 | NP_079229.2 | ||
| XM_005253938.1 | XP_005253995.1 | |||
| XM_006719606.2 | XP_006719669.1 | |||
| XM_017019977.1 | XP_016875466.1 | |||
| AB054990.1 | BAC80174.1 | |||
| AK023151.1 | BAB14432.1 | |||
| AK292485.1 | ||||
| AK309772.1 | ||||
| AL832394.1 | ||||
| BC113585.1 | ||||
| BC113587.1 | ||||
| BX641140.1 | CAE46062.1 | |||
| BX647450.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv52789 | Chr.12:112034771 - 112038935 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map