Assay Details
Target Gene Details
Entrez Gene ID: | 56994 |
Gene Name: | choline phosphotransferase 1 |
Gene Aliases: |
CPT, CPT1 |
Location: |
Chr.12:101697639-101752038 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHPT1 | NM_020244.2 | NP_064629.2 | ||
| XM_011538574.1 | XP_011536876.1 | |||
| AF047431.1 | AAD44019.1 | |||
| AF195623.1 | AAF87947.1 | |||
| AF195624.1 | AAF87948.1 | |||
| AK075211.1 | ||||
| AK093067.1 | ||||
| AK226127.1 | ||||
| BC020819.1 | AAH20819.1 | |||
| BC050429.1 | AAH50429.1 | |||
| CD300296.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832497 | Chr.12:101667198 - 101861862 on Build GRCh38 | Gain |
|
| dgv87e215 | Chr.12:101708223 - 101713722 on Build GRCh38 | Deletion |
|
| nsv498804 | Chr.12:101708628 - 101713313 on Build GRCh38 | Loss |
|
| esv21962 | Chr.12:101708956 - 101713345 on Build GRCh38 | Loss |
|
| nsv435634 | Chr.12:101707835 - 101713695 on Build GRCh38 | Deletion |
|
| nsv560007 | Chr.12:101707848 - 101713222 on Build GRCh38 | Loss |
|
| nsv1069479 | Chr.12:101708621 - 101713322 on Build GRCh38 | Deletion |
|
| esv3630579 | Chr.12:101708632 - 101713337 on Build GRCh38 | Loss |
|
| esv9053 | Chr.12:101708428 - 101713516 on Build GRCh38 | Loss |
|
| nsv859 | Chr.12:101696419 - 101720084 on Build GRCh38 | Deletion |
|
| nsv560006 | Chr.12:101703234 - 101714567 on Build GRCh38 | Loss |
|
| nsv826502 | Chr.12:101708728 - 101712123 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map