Assay Details
Target Gene Details
Entrez Gene ID: | 57460 |
Gene Name: | protein phosphatase, Mg2+/Mn2+ dependent 1H |
Gene Aliases: |
ARHCL1, NERPP-2C, URCC2 |
Location: |
Chr.12:62643982-62935135 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPM1H | NM_020700.1 | NP_065751.1 | ||
| XM_011538578.2 | XP_011536880.1 | |||
| XM_011538579.1 | XP_011536881.1 | |||
| AB032983.1 | BAA86471.1 | |||
| AB084258.1 | ||||
| AK023315.1 | ||||
| BC157843.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv741 | Chr.12:62644475 - 62671066 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115316642] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map