Assay Details
Target Gene Details
Entrez Gene ID: | 339 |
Gene Name: | apolipoprotein B mRNA editing enzyme catalytic subunit 1 |
Gene Aliases: |
APOBEC-1, BEDP, CDAR1, HEPR |
Location: |
Chr.12:7649400-7670599 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APOBEC1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521241 | Chr.12:7663155 - 7673709 on Build GRCh38 | Loss |
|
| dgv1350n100 | Chr.12:7502541 - 7672399 on Build GRCh38 | Loss |
|
| nsv826213 | Chr.12:7578336 - 7678048 on Build GRCh38 | Gain |
|
| nsv557273 | Chr.12:7621432 - 7731755 on Build GRCh38 | Loss |
|
| nsv557272 | Chr.12:7604110 - 7669096 on Build GRCh38 | Loss |
|
| dgv5e196 | Chr.12:7621924 - 7954221 on Build GRCh38 | Duplication |
|

Genomic Map