Assay Details
Target Gene Details
Entrez Gene ID: | 51564 |
Gene Name: | histone deacetylase 7 |
Gene Aliases: |
HD7, HD7A, HDAC7A |
Location: |
Chr.12:47782710-47820612 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HDAC7 | NM_001098416.3 | NP_001091886.1 | ||
| NM_001308090.1 | NP_001295019.1 | |||
| NM_015401.4 | NP_056216.2 | |||
| XM_011538478.1 | XP_011536780.1 | |||
| XM_011538479.1 | XP_011536781.1 | |||
| XM_011538480.1 | XP_011536782.1 | |||
| XM_011538481.1 | XP_011536783.1 | |||
| XM_011538482.1 | XP_011536784.1 | |||
| XM_011538483.1 | XP_011536785.1 | |||
| XM_017019455.1 | XP_016874944.1 | |||
| XM_017019456.1 | XP_016874945.1 | |||
| AF239243.1 | AAF63491.1 | |||
| AK001190.1 | BAA91545.1 | |||
| AK024469.1 | BAB15759.1 | |||
| AK225910.1 | ||||
| AK299292.1 | ||||
| AK301545.1 | ||||
| AK303481.1 | ||||
| AL117455.1 | CAB55935.1 | |||
| AY302468.1 | AAQ18232.1 | |||
| AY321367.1 | AAP84704.1 | |||
| BC064840.1 | AAH64840.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv94n21 | Chr.12:47725779 - 47821397 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map