Assay Details
Target Gene Details
Entrez Gene ID: | 7347 |
Gene Name: | ubiquitin C-terminal hydrolase L3 |
Gene Aliases: |
UCH-L3 |
Location: |
Chr.13:75549480-75606020 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UCHL3 | NM_001270952.1 | NP_001257881.1 | ||
| NM_006002.4 | NP_005993.1 | |||
| XM_011535212.1 | XP_011533514.1 | |||
| XM_011535213.2 | XP_011533515.1 | |||
| XM_011535214.2 | XP_011533516.1 | |||
| XM_017020725.1 | XP_016876214.1 | |||
| XM_017020726.1 | XP_016876215.1 | |||
| XM_017020727.1 | XP_016876216.1 | |||
| AK313665.1 | ||||
| BC018125.2 | AAH18125.1 | |||
| BQ427804.1 | ||||
| BT019359.1 | AAV38166.1 | |||
| CR456855.1 | CAG33136.1 | |||
| M30496.1 | AAA36791.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2751152 | Chr.13:75570063 - 75714282 on Build GRCh38 | Loss |
|
| nsv521444 | Chr.13:74139302 - 75679218 on Build GRCh38 | Gain |
|
| esv2761803 | Chr.13:75369173 - 75587638 on Build GRCh38 | Gain |
|
| nsv562347 | Chr.13:75558104 - 75579279 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs145090403] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map