Assay Details
Target Gene Details
Entrez Gene ID: | 23077 |
Gene Name: | MYC binding protein 2, E3 ubiquitin protein ligase |
Gene Aliases: |
Myc-bp2, PAM, Phr |
Location: |
Chr.13:77044657-77327066 on Build GRCh38 |
Assay Gene Location: | Within Intron 82 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832653 | Chr.13:76948899 - 77115221 on Build GRCh38 | Loss |
|
| esv2759951 | Chr.13:77050897 - 77194329 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map