Assay Details
Target Gene Details
Entrez Gene ID: | 6011 |
Gene Name: | G protein-coupled receptor kinase 1 |
Gene Aliases: |
GPRK1, RHOK, RK |
Location: |
Chr.13:113645771-113735664 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRK1 | NM_002929.2 | NP_002920.1 | ||
| XM_017020684.1 | XP_016876173.1 | |||
| AK289912.1 | ||||
| BC140945.1 | ||||
| L77503.1 | AAG50439.1 | |||
| U63973.1 | AAB05929.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2748349 | Chr.13:113670392 - 113670927 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map