Assay Details
Target Gene Details
Entrez Gene ID: | 6011 |
Gene Name: | G protein-coupled receptor kinase 1 |
Gene Aliases: |
GPRK1, RHOK, RK |
Location: |
Chr.13:113645771-113735664 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRK1 | XM_017020684.1 | XP_016876173.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832726 | Chr.13:113559093 - 113663088 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map