Assay Details
Target Gene Details
Entrez Gene ID: | 2262 |
Gene Name: | glypican 5 |
Gene Aliases: |
- |
Location: |
Chr.13:91398619-92867237 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPC5 | NM_004466.5 | NP_004457.1 | ||
| XM_017020435.1 | XP_016875924.1 | |||
| AF001462.1 | AAC12261.1 | |||
| AK312815.1 | ||||
| BC039730.1 | AAH39730.1 | |||
| U66033.1 | AAC51118.1 |
Target Gene Details
Entrez Gene ID: | 100873970 |
Gene Name: | GPC5 antisense RNA 2 |
Gene Aliases: |
- |
Location: |
Chr.13:92339794-92352207 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPC5-AS2 | NR_120382.1 | 1 | 73 | |
| AI827689.1 | 3 | 341 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3582694 | Chr.13:91877905 - 93587318 on Build GRCh38 | Gain |
|
| nsv1122 | Chr.13:92343416 - 92374054 on Build GRCh38 | Insertion |
|
| dgv1730n100 | Chr.13:92300848 - 92399755 on Build GRCh38 | Gain |
|
| nsv562727 | Chr.13:92308898 - 92386660 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map