Assay Details
Target Gene Details
Entrez Gene ID: | 8100 |
Gene Name: | intraflagellar transport 88 |
Gene Aliases: |
D13S1056E, DAF19, TG737, TTC10, hTg737 |
Location: |
Chr.13:20566446-20691437 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759925 | Chr.13:20398583 - 20650339 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map