Assay Details
Target Gene Details
Entrez Gene ID: | 6011 |
Gene Name: | G protein-coupled receptor kinase 1 |
Gene Aliases: |
GPRK1, RHOK, RK |
Location: |
Chr.13:113645771-113735664 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRK1 | XM_017020684.1 | XP_016876173.1 |
More Information
Set Membership: |
|

Genomic Map