Assay Details
Target Gene Details
Entrez Gene ID: | 4008 |
Gene Name: | LIM domain 7 |
Gene Aliases: |
FBX20, FBXO20, LOMP |
Location: |
Chr.13:75620434-75859870 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LMO7 | NM_001306080.1 | NP_001293009.1 | ||
| NM_005358.5 | NP_005349.3 | |||
| AF330045.1 | AAL37480.1 | |||
| AK092052.1 | ||||
| DA313248.1 | ||||
| FJ711162.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2751152 | Chr.13:75570063 - 75714282 on Build GRCh38 | Loss |
|
| nsv521444 | Chr.13:74139302 - 75679218 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115299350] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map