Assay Details
Target Gene Details
Entrez Gene ID: | 730249 |
Gene Name: | aconitate decarboxylase 1 |
Gene Aliases: |
CAD, IRG1 |
Location: |
Chr.13:76948559-76958642 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACOD1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832653 | Chr.13:76948899 - 77115221 on Build GRCh38 | Loss |
|
| nsv832651 | Chr.13:76788718 - 76974704 on Build GRCh38 | Gain |
|
| nsv832652 | Chr.13:76855107 - 77041507 on Build GRCh38 | Gain |
|

Genomic Map