Assay Details
Target Gene Details
Entrez Gene ID: | 100507533 |
Gene Name: | SOX21 antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.13:94712716-94715945 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOX21-AS1 | NR_046514.1 | 1 | 108 | |
| AK055459.1 | 1 | 110 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1070605 | Chr.13:94711245 - 94714046 on Build GRCh38 | Deletion |
|
| nsv832686 | Chr.13:94554488 - 94748738 on Build GRCh38 | Gain |
|
| esv28079 | Chr.13:94710850 - 94713584 on Build GRCh38 | Loss |
|
| esv34004 | Chr.13:94254283 - 94740390 on Build GRCh38 | Loss |
|
| nsv562766 | Chr.13:94712121 - 94713517 on Build GRCh38 | Loss |
|
| nsv826750 | Chr.13:94706248 - 94715816 on Build GRCh38 | Gain |
|
| dgv3325n54 | Chr.13:94711363 - 94713127 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs536137591] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map