Assay Details
Target Gene Details
Entrez Gene ID: | 1241 |
Gene Name: | leukotriene B4 receptor |
Gene Aliases: |
BLT1, BLTR, CMKRL1, GPR16, LTB4R1, LTBR1, P2RY7, P2Y7 |
Location: |
Chr.14:24311499-24318036 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LTB4R | NM_001143919.2 | NP_001137391.1 | ||
| D89079.1 | BAA20424.1 | |||
| U41070.1 | AAC50628.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564058 | Chr.14:24302079 - 24317770 on Build GRCh38 | Loss |
|
| nsv564060 | Chr.14:24307195 - 24340589 on Build GRCh38 | Loss |
|
| esv2669637 | Chr.14:24274158 - 24318169 on Build GRCh38 | Deletion |
|
| esv3633804 | Chr.14:24274134 - 24318147 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map