Assay Details
Target Gene Details
Entrez Gene ID: | 55775 |
Gene Name: | tyrosyl-DNA phosphodiesterase 1 |
Gene Aliases: |
- |
Location: |
Chr.14:89954935-90044764 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TDP1 | NM_001008744.1 | NP_001008744.1 | ||
| NM_018319.3 | NP_060789.2 | |||
| XM_005267847.2 | XP_005267904.1 | |||
| XM_005267848.2 | XP_005267905.1 | |||
| XM_006720197.3 | XP_006720260.1 | |||
| XM_006720199.1 | XP_006720262.1 | |||
| XM_011536942.2 | XP_011535244.1 | |||
| XM_011536944.1 | XP_011535246.1 | |||
| XM_017021439.1 | XP_016876928.1 | |||
| XM_017021440.1 | XP_016876929.1 | |||
| XM_017021441.1 | XP_016876930.1 | |||
| XM_017021443.1 | XP_016876932.1 | |||
| XM_017021444.1 | XP_016876933.1 | |||
| XM_017021446.1 | XP_016876935.1 | |||
| AK001952.1 | BAA91997.1 | |||
| AK093235.1 | ||||
| AK315549.1 | ||||
| BC006083.1 | AAH06083.1 | |||
| BC015474.1 | AAH15474.1 | |||
| BC107877.1 | AAI07878.1 | |||
| BU849335.1 | ||||
| BX161451.1 | CAD61915.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832854 | Chr.14:89877925 - 90070680 on Build GRCh38 | Gain |
|
| nsv832853 | Chr.14:89818942 - 89990332 on Build GRCh38 | Gain |
|
| nsv1045864 | Chr.14:89882005 - 89968468 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map