Assay Details
Target Gene Details
Entrez Gene ID: | 54207 |
Gene Name: | potassium two pore domain channel subfamily K member 10 |
Gene Aliases: |
K2p10.1, PPP1R97, TREK-2, TREK2 |
Location: |
Chr.14:88180108-88326912 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNK10 | NM_021161.4 | NP_066984.1 | ||
| NM_138317.2 | NP_612190.1 | |||
| NM_138318.2 | NP_612191.1 | |||
| XM_011536840.2 | XP_011535142.1 | |||
| AF279890.1 | AAG15191.1 | |||
| AF385399.1 | AAL95705.1 | |||
| AF385400.1 | AAL95706.1 | |||
| AK290518.1 | ||||
| AK313499.1 | ||||
| AK315263.1 | ||||
| BC075021.2 | AAH75021.1 | |||
| BC075022.2 | AAH75022.1 | |||
| EU978938.1 | ||||
| EU978939.1 | ||||
| EU978940.1 | ||||
| EU978941.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv85763 | Chr.14:88195040 - 88197572 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map