Assay Details
Target Gene Details
Entrez Gene ID: | 55632 |
Gene Name: | G2/M-phase specific E3 ubiquitin protein ligase |
Gene Aliases: |
KIAA1333, PHF7B |
Location: |
Chr.14:30559123-30620064 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| G2E3 | NM_001308097.1 | NP_001295026.1 | ||
| NM_017769.4 | NP_060239.2 | |||
| XM_011536926.1 | XP_011535228.1 | |||
| AB037754.1 | ||||
| AK000340.1 | BAA91095.1 | |||
| AK093536.1 | ||||
| AK225346.1 | ||||
| AK295571.1 | ||||
| AK296872.1 | ||||
| BC000973.2 | AAH00973.2 | |||
| DA491192.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564169 | Chr.14:30349881 - 31072288 on Build GRCh38 | Gain |
|
| nsv526536 | Chr.14:28322483 - 30669172 on Build GRCh38 | Gain |
|
| esv33189 | Chr.14:30331017 - 31442539 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map