Assay Details
Target Gene Details
Entrez Gene ID: | 123096 |
Gene Name: | solute carrier family 25 member 29 |
Gene Aliases: |
C14orf69, CACL, ORNT3 |
Location: |
Chr.14:100277986-100306547 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A29 | NM_001039355.2 | NP_001034444.1 | ||
| NM_001291813.1 | NP_001278742.1 | |||
| NM_001291814.1 | NP_001278743.1 | |||
| NM_152333.3 | NP_689546.1 | |||
| XM_006720038.1 | XP_006720101.1 | |||
| XM_006720039.3 | XP_006720102.1 | |||
| XM_011536443.1 | XP_011534745.1 | |||
| XM_011536444.1 | XP_011534746.1 | |||
| XM_011536446.1 | XP_011534748.1 | |||
| XM_011536447.1 | XP_011534749.1 | |||
| XM_011536448.1 | XP_011534750.1 | |||
| XM_017020979.1 | XP_016876468.1 | |||
| XM_017020980.1 | 4 | 4315 | XP_016876469.1 | |
| XM_017020981.1 | 3 | 3839 | XP_016876470.1 | |
| XM_017020982.1 | 3 | 2369 | XP_016876471.1 | |
| XM_017020983.1 | 3 | 2359 | XP_016876472.1 | |
| XM_017020984.1 | 3 | 2073 | XP_016876473.1 | |
| XM_017020985.1 | 4 | 2118 | XP_016876474.1 | |
| XM_017020986.1 | 3 | 1554 | XP_016876475.1 | |
| XM_017020987.1 | 4 | 1534 | XP_016876476.1 | |
| AK096294.1 | BAC04751.1 | |||
| AK123821.1 | ||||
| AK172788.1 | BAD18767.1 | |||
| AK289627.1 | ||||
| AL832299.1 | 1 | 1536 | ||
| BI828620.1 | ||||
| BX247983.1 | CAD62317.1 | |||
| DA201175.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3635502 | Chr.14:100275457 - 100309132 on Build GRCh38 | Gain |
|
| nsv832875 | Chr.14:100192592 - 100368905 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map