Assay Details
Target Gene Details
Entrez Gene ID: | 123096 |
Gene Name: | solute carrier family 25 member 29 |
Gene Aliases: |
C14orf69, CACL, ORNT3 |
Location: |
Chr.14:100277986-100306547 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A29 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3635496 | Chr.14:100169585 - 100290965 on Build GRCh38 | Gain |
|
| esv3635502 | Chr.14:100275457 - 100309132 on Build GRCh38 | Gain |
|
| nsv565781 | Chr.14:100287405 - 100293628 on Build GRCh38 | Loss |
|
| nsv832875 | Chr.14:100192592 - 100368905 on Build GRCh38 | Loss |
|

Genomic Map