Assay Details
Target Gene Details
Entrez Gene ID: | 113146 |
Gene Name: | AHNAK nucleoprotein 2 |
Gene Aliases: |
C14orf78 |
Location: |
Chr.14:104937253-104978357 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AHNAK2 | NM_138420.2 | 7 | 1473 | NP_612429.2 |
| XM_005267299.1 | 7 | 1326 | XP_005267356.1 | |
| AK094143.1 | 7 | 1326 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1438 | Chr.14:104946391 - 104991244 on Build GRCh38 | Deletion |
|
| nsv566141 | Chr.14:104938047 - 105089059 on Build GRCh38 | Gain |
|
| esv3635629 | Chr.14:104942355 - 104955280 on Build GRCh38 | Gain |
|
| nsv1049253 | Chr.14:104918622 - 105005518 on Build GRCh38 | Gain |
|
| nsv952276 | Chr.14:104922864 - 105021963 on Build GRCh38 | Deletion |
|
| nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map