Assay Details
Target Gene Details
Entrez Gene ID: | 54207 |
Gene Name: | potassium two pore domain channel subfamily K member 10 |
Gene Aliases: |
K2p10.1, PPP1R97, TREK-2, TREK2 |
Location: |
Chr.14:88180108-88326912 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNK10 | NM_021161.4 | 7 | 5728 | NP_066984.1 |
| NM_138317.2 | 7 | 5737 | NP_612190.1 | |
| NM_138318.2 | 7 | 5408 | NP_612191.1 | |
| XM_011536840.2 | 6 | 5113 | XP_011535142.1 | |
| AK125389.1 | 1 | 1159 |
More Information
Set Membership: |
|

Genomic Map