Assay Details
Target Gene Details
Entrez Gene ID: | 23508 |
Gene Name: | tetratricopeptide repeat domain 9 |
Gene Aliases: |
TTC9A |
Location: |
Chr.14:70641787-70675360 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC9 | NM_015351.1 | NP_056166.1 | ||
| BC047950.1 | AAH47950.1 | |||
| BX407741.2 | ||||
| D86980.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2672447 | Chr.14:70655182 - 70657801 on Build GRCh38 | Deletion |
|
| esv3634866 | Chr.14:70655178 - 70657865 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map