Assay Details
Target Gene Details
Entrez Gene ID: | 9369 |
Gene Name: | neurexin 3 |
Gene Aliases: |
C14orf60 |
Location: |
Chr.14:77979904-79868290 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NRXN3 | XM_017021790.1 | XP_016877279.1 | ||
| XM_017021791.1 | XP_016877280.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34345 | Chr.14:77904112 - 78030244 on Build GRCh38 | Loss |
|
| esv2759999 | Chr.14:77912813 - 78035676 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map