Assay Details
Target Gene Details
Entrez Gene ID: | 23508 |
Gene Name: | tetratricopeptide repeat domain 9 |
Gene Aliases: |
TTC9A |
Location: |
Chr.14:70641787-70675360 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC9 | NM_015351.1 | 1 | 583 | NP_056166.1 |
| BC047950.1 | 1 | 405 | AAH47950.1 | |
| BX407741.2 | 1 | 409 | ||
| D86980.1 | 1 | 583 |
More Information
Set Membership: |
|

Genomic Map