Assay Details
Target Gene Details
Entrez Gene ID: | 23269 |
Gene Name: | MGA, MAX dimerization protein |
Gene Aliases: |
MAD5, MXD5 |
Location: |
Chr.15:41621139-41769943 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MGA | XM_005254246.2 | XP_005254303.1 | ||
| XM_005254247.2 | XP_005254304.1 | |||
| XM_006720445.3 | XP_006720508.1 | |||
| XM_017022029.1 | XP_016877518.1 | |||
| BC038449.2 | AAH38449.1 |
More Information
Set Membership: |
|

Genomic Map