Assay Details
Target Gene Details
Entrez Gene ID: | 9915 |
Gene Name: | aryl hydrocarbon receptor nuclear translocator 2 |
Gene Aliases: |
WEDAS, bHLHe1 |
Location: |
Chr.15:80404350-80597937 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARNT2 | NM_014862.3 | NP_055677.3 | ||
| AB002305.1 | BAA20766.2 | |||
| AK295763.1 | ||||
| BC036099.1 | AAH36099.1 | |||
| BX538026.1 | CAD97972.1 | |||
| BX647212.1 | ||||
| KJ534772.1 | ||||
| KJ534773.1 | ||||
| KJ534986.1 | ||||
| KJ534995.1 | ||||
| KJ534996.1 | ||||
| KJ535008.1 | ||||
| KJ535009.1 | ||||
| KJ535057.1 |
Target Gene Details
Entrez Gene ID: | 101929586 |
Gene Name: | uncharacterized LOC101929586 |
Gene Aliases: |
- |
Location: |
Chr.15:80554609-80562944 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929586 | NR_120363.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471685 | Chr.15:80530860 - 80711048 on Build GRCh38 | Gain |
|
| nsv570235 | Chr.15:80541802 - 80598985 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map