Assay Details
Target Gene Details
Entrez Gene ID: | 645212 |
Gene Name: | EIF3J antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.15:44534505-44536923 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF3J-AS1 | NR_034170.1 | 1 | 660 | |
| NR_034171.1 | 2 | 195 | ||
| AK092547.1 | 1 | 411 | ||
| BC026043.1 | 1 | 648 | AAH26043.1 | |
| DB480720.1 | 2 | 195 |

Genomic Map