Assay Details
Target Gene Details
Entrez Gene ID: | 8826 |
Gene Name: | IQ motif containing GTPase activating protein 1 |
Gene Aliases: |
HUMORFA01, SAR1, p195 |
Location: |
Chr.15:90388241-90502243 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IQGAP1 | NM_003870.3 | NP_003861.1 | ||
| AK304337.1 | ||||
| AK307643.1 | ||||
| BC005906.1 | ||||
| BC020549.1 | AAH20549.1 | |||
| BC037425.1 | ||||
| BC064971.1 | AAH64971.1 | |||
| BC089385.1 | AAH89385.1 | |||
| BC110811.1 | AAI10812.1 | |||
| BC139731.1 | ||||
| BC151834.1 | ||||
| D29640.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833087 | Chr.15:90208675 - 90425725 on Build GRCh38 | Loss |
|
| nsv457246 | Chr.15:90282201 - 90455896 on Build GRCh38 | Gain |
|
| esv2751541 | Chr.15:90245652 - 90457213 on Build GRCh38 | Gain |
|
| esv2761843 | Chr.15:90289439 - 90462457 on Build GRCh38 | Gain |
|
| nsv952027 | Chr.15:90394769 - 90414268 on Build GRCh38 | Duplication |
|
| esv3637227 | Chr.15:90409864 - 90417474 on Build GRCh38 | Loss |
|
| dgv2659n100 | Chr.15:90301813 - 90415770 on Build GRCh38 | Gain |
|
| dgv2658n100 | Chr.15:90290495 - 90466149 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map