Assay Details
Target Gene Details
Entrez Gene ID: | 145853 |
Gene Name: | chromosome 15 open reading frame 61 |
Gene Aliases: |
- |
Location: |
Chr.15:67521184-67527303 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C15orf61 | NM_001143936.1 | 1 | 260 | NP_001137408.1 |
| AK294012.1 | 1 | 195 | ||
| AY726587.1 | 1 | 152 |
Target Gene Details
Entrez Gene ID: | 100506686 |
Gene Name: | IQCH antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.15:67403611-67521844 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IQCH-AS1 | NR_040051.1 | |||
| NR_040052.1 | ||||
| NR_040054.1 | ||||
| AL598736.1 | ||||
| BX332831.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv569799 | Chr.15:67511204 - 67585107 on Build GRCh38 | Loss |
|
| esv3636789 | Chr.15:67514940 - 67521624 on Build GRCh38 | Loss |
|

Genomic Map