Assay Details
Target Gene Details
Entrez Gene ID: | 22801 |
Gene Name: | integrin subunit alpha 11 |
Gene Aliases: |
HsT18964 |
Location: |
Chr.15:68297433-68432312 on Build GRCh38 |
Assay Gene Location: | Within Intron 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITGA11 | NM_001004439.1 | NP_001004439.1 | ||
| XM_005254228.3 | XP_005254285.1 | |||
| XM_011521363.2 | XP_011519665.1 | |||
| AF109681.1 | AAF01258.1 | |||
| AF111799.1 | AAL39001.1 | |||
| AF137378.2 | AAD51919.2 | |||
| AK095863.1 | ||||
| AK292445.1 | ||||
| AK304778.1 | ||||
| AL359064.1 | CAB94392.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33024 | Chr.15:68299521 - 68336679 on Build GRCh38 | Gain |
|
| esv2760036 | Chr.15:68245734 - 68522324 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map