Assay Details
Target Gene Details
Entrez Gene ID: | 5151 |
Gene Name: | phosphodiesterase 8A |
Gene Aliases: |
HsT19550 |
Location: |
Chr.15:84980513-85139145 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDE8A | NM_001243137.1 | NP_001230066.1 | ||
| NM_002605.2 | NP_002596.1 | |||
| NM_173454.1 | NP_775656.1 | |||
| XM_017022308.1 | XP_016877797.1 | |||
| XM_017022309.1 | XP_016877798.1 | |||
| XM_017022310.1 | XP_016877799.1 | |||
| XM_017022311.1 | XP_016877800.1 | |||
| AF332653.1 | AAK57641.1 | |||
| AF388183.1 | AAL18610.1 | |||
| AF388184.1 | AAL18611.1 | |||
| AF388185.1 | AAL18612.1 | |||
| AF388186.1 | AAL18613.1 | |||
| AF388187.1 | AAL18614.1 | |||
| AK074280.1 | ||||
| BC060762.1 | AAH60762.1 | |||
| BC075822.1 | AAH75822.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1040994 | Chr.15:84992252 - 85041926 on Build GRCh38 | Gain |
|
| esv2672879 | Chr.15:84183834 - 85188520 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map