Assay Details
Target Gene Details
Entrez Gene ID: | 100130855 |
Gene Name: | USP3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.15:63587710-63600827 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP3-AS1 | NR_034080.1 | |||
| AK093077.1 | ||||
| DB444752.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833034 | Chr.15:63500671 - 63657512 on Build GRCh38 | Loss |
|

Genomic Map