Assay Details
Target Gene Details
Entrez Gene ID: | 10519 |
Gene Name: | calcium and integrin binding 1 |
Gene Aliases: |
CIB, CIBP, KIP1, PRKDCIP, SIP2-28 |
Location: |
Chr.15:90230245-90265759 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CIB1 | NM_001277764.1 | NP_001264693.1 | ||
| NM_006384.3 | NP_006375.2 | |||
| NR_102427.1 | ||||
| NR_102428.1 | ||||
| XM_006720375.1 | XP_006720438.1 | |||
| AA769426.1 | ||||
| AB451276.1 | ||||
| AB451406.1 | ||||
| BC000846.2 | AAH00846.1 | |||
| BG231771.1 | ||||
| BG759961.1 | ||||
| BI520476.1 | ||||
| CD622547.1 | ||||
| CR456955.1 | CAG33236.1 | |||
| JQ246073.1 | ||||
| U82226.1 | AAC51106.1 | |||
| U83236.1 | AAB39758.1 | |||
| U85611.1 | AAB53387.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3637214 | Chr.15:90184741 - 90235676 on Build GRCh38 | Gain |
|
| nsv833087 | Chr.15:90208675 - 90425725 on Build GRCh38 | Loss |
|
| nsv1052652 | Chr.15:90101186 - 90298961 on Build GRCh38 | Gain |
|
| dgv112e55 | Chr.15:90079272 - 90249274 on Build GRCh38 | Gain |
|
| esv2760054 | Chr.15:90022724 - 90266401 on Build GRCh38 | Gain |
|
| nsv817707 | Chr.15:90104624 - 90241698 on Build GRCh38 | Gain |
|
| nsv442384 | Chr.15:90089378 - 90235161 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map