Assay Details
Target Gene Details
Entrez Gene ID: | 9960 |
Gene Name: | ubiquitin specific peptidase 3 |
Gene Aliases: |
SIH003, UBP |
Location: |
Chr.15:63504511-63594640 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP3 | NM_001256702.1 | 14 | 3149 | NP_001243631.1 |
| NM_006537.3 | 15 | 3281 | NP_006528.2 | |
| NR_046341.1 | 16 | 3434 | ||
| NR_046342.1 | 16 | 3397 | ||
| XM_017022763.1 | 17 | 4280 | XP_016878252.1 | |
| XM_017022764.1 | 16 | 4216 | XP_016878253.1 | |
| XM_017022765.1 | 16 | 3422 | XP_016878254.1 | |
| XM_017022766.1 | 17 | 3465 | XP_016878255.1 | |
| XM_017022767.1 | 18 | 4396 | XP_016878256.1 | |
| XM_017022768.1 | 18 | 4470 | XP_016878257.1 | |
| AK094444.1 | 1 | 400 | ||
| BX647760.1 | 16 | 3175 |
Target Gene Details
Entrez Gene ID: | 100130855 |
Gene Name: | USP3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.15:63587710-63600827 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP3-AS1 | NR_034080.1 | |||
| AK093077.1 | ||||
| DB444752.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833034 | Chr.15:63500671 - 63657512 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76115381] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map