Assay Details
Target Gene Details
Entrez Gene ID: | 83986 |
Gene Name: | family with sequence similarity 234 member A |
Gene Aliases: |
C16orf9, ITFG3, gs19 |
Location: |
Chr.16:234546-269943 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM234A | NM_001284497.1 | NP_001271426.1 | ||
| NM_032039.3 | NP_114428.1 | |||
| NR_104317.1 | ||||
| XM_005255622.1 | XP_005255679.1 | |||
| XM_005255623.2 | XP_005255680.1 | |||
| XM_006720957.1 | XP_006721020.1 | |||
| XM_011522691.1 | XP_011520993.1 | |||
| XM_011522692.1 | XP_011520994.1 | |||
| XM_017023760.1 | XP_016879249.1 | |||
| XM_017023761.1 | XP_016879250.1 | |||
| XM_017023762.1 | XP_016879251.1 | |||
| XM_017023763.1 | XP_016879252.1 | |||
| XM_017023764.1 | XP_016879253.1 | |||
| XM_017023765.1 | XP_016879254.1 | |||
| AK057165.1 | BAB71374.1 | |||
| AK294581.1 | ||||
| AK310766.1 | ||||
| AL136542.1 | CAB66477.1 | |||
| CN262982.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518291 | Chr.16:47354 - 254515 on Build GRCh38 | Loss |
|
| esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication |
|
| nsv1044142 | Chr.16:10765 - 255644 on Build GRCh38 | Gain |
|
| esv3570031 | Chr.16:243389 - 278107 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map