Assay Details
Target Gene Details
Entrez Gene ID: | 101927102 |
Gene Name: | ITFG1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.16:47144068-47162747 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITFG1-AS1 | NR_110903.1 | |||
| NR_110904.1 | ||||
| BE466653.1 | ||||
| BF115500.1 | ||||
| BI462694.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1066115 | Chr.16:46429858 - 47284504 on Build GRCh38 | Gain |
|
| nsv528423 | Chr.16:47129963 - 47313869 on Build GRCh38 | Loss |
|

Genomic Map