Assay Details
Target Gene Details
Entrez Gene ID: | 3801 |
Gene Name: | kinesin family member C3 |
Gene Aliases: |
- |
Location: |
Chr.16:57758217-57863053 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIFC3 | XM_005255937.1 | XP_005255994.1 | ||
| XM_006721188.1 | XP_006721251.1 | |||
| XM_011523075.1 | XP_011521377.1 | |||
| XM_011523076.1 | XP_011521378.1 | |||
| XM_017023221.1 | XP_016878710.1 | |||
| AK295926.1 | ||||
| DQ426879.1 |
Target Gene Details
Entrez Gene ID: | 388282 |
Gene Name: | uncharacterized LOC388282 |
Gene Aliases: |
- |
Location: |
Chr.16:57809426-57817114 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC388282 | NM_001278081.1 | 2 | 280 | NP_001265010.1 |
| XM_011523082.2 | 2 | 1491 | XP_011521384.1 | |
| AF452719.1 | AAP13354.1 | |||
| BG775641.1 | 2 | 281 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525464 | Chr.16:57714251 - 57832787 on Build GRCh38 | Gain |
|
| nsv509622 | Chr.16:57720104 - 57826484 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map