Assay Details
Target Gene Details
Entrez Gene ID: | 54971 |
Gene Name: | BTG3 associated nuclear protein |
Gene Aliases: |
BEND1, SMAR1, SMARBP1 |
Location: |
Chr.16:87949216-88077318 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1059786 | Chr.16:87811419 - 88019505 on Build GRCh38 | Gain |
|
| nsv1919 | Chr.16:87963774 - 87979608 on Build GRCh38 | Deletion |
|
| esv3892935 | Chr.16:87808912 - 88010269 on Build GRCh38 | Loss |
|
| esv2758668 | Chr.16:87885744 - 88080126 on Build GRCh38 | Loss |
|
| nsv1066924 | Chr.16:87911850 - 87998507 on Build GRCh38 | Loss |
|
| nsv1062946 | Chr.16:87635801 - 88019505 on Build GRCh38 | Gain |
|
| dgv36n68 | Chr.16:87801736 - 87983550 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73240802] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map