Assay Details
Target Gene Details
Entrez Gene ID: | 23199 |
Gene Name: | Gse1 coiled-coil protein |
Gene Aliases: |
CRHSP24, KIAA0182 |
Location: |
Chr.16:85169512-85676206 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GSE1 | XM_005255859.4 | 1 | 1560 | XP_005255916.3 |
| XM_005255860.3 | 1 | 1567 | XP_005255917.3 | |
| XM_005255861.4 | 1 | 1560 | XP_005255918.3 | |
| XM_017023084.1 | 1 | 1732 | XP_016878573.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833312 | Chr.16:85158268 - 85327826 on Build GRCh38 | Loss |
|
| nsv827785 | Chr.16:85168227 - 85171735 on Build GRCh38 | Gain |
|
| dgv3059n100 | Chr.16:85068826 - 85219999 on Build GRCh38 | Gain |
|
| nsv483044 | Chr.16:84166395 - 87066394 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map