Assay Details
Target Gene Details
Entrez Gene ID: | 9810 |
Gene Name: | ring finger protein 40 |
Gene Aliases: |
BRE1B, RBP95, STARING |
Location: |
Chr.16:30761612-30800231 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF40 | NM_001207033.1 | NP_001193962.1 | ||
| NM_001207034.1 | NP_001193963.1 | |||
| NM_001286572.2 | NP_001273501.1 | |||
| NM_014771.3 | NP_055586.1 | |||
| XM_011545997.1 | XP_011544299.1 | |||
| AB014561.1 | ||||
| AF122819.1 | AAG13723.1 | |||
| AK027406.1 | BAB55092.1 | |||
| AK054773.1 | 1 | 662 | ||
| AK075526.1 | ||||
| AK131310.1 | BAD18476.1 | |||
| AK291666.1 | ||||
| AK293710.1 | ||||
| AK294258.1 | ||||
| AK297531.1 | ||||
| AK304521.1 | ||||
| BC004527.2 | AAH04527.2 | |||
| BC006133.1 | AAH06133.1 | |||
| BC011769.2 | AAH11769.1 | |||
| BC018647.2 | AAH18647.1 | |||
| BC030802.1 | AAH30802.2 | |||
| BX640763.1 | CAE45869.1 | |||
| CR627431.1 | CAH10518.1 | |||
| KU178492.1 | ||||
| KU178493.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469724 | Chr.16:30621124 - 30828942 on Build GRCh38 | Loss |
|
| nsv833188 | Chr.16:30621125 - 30828942 on Build GRCh38 | Loss |
|
| nsv9438 | Chr.16:30758882 - 30788121 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map